missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Monobutyl Itaconate, TCI America™

$106.95 - $748.63

Chemical Identifiers

CAS 6439-57-2
Molecular Formula C9H13O4-
Molecular Weight (g/mol) 185.199
MDL Number MFCD00046533
InChI Key DYYFCJRYZVHEKQ-UHFFFAOYSA-M
Synonym beta-Butyl Itaconate, 4-Butyl Methylenesuccinate, Itaconic Acid Monobutyl Ester
PubChem CID 21225725
IUPAC Name 3-butoxycarbonylbut-3-enoate
SMILES CCCCOC(=O)C(=C)CC(=O)[O-]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
I026725G View Documents TCI America
I026725G
25g Each for $106.95
 
I0267500G View Documents TCI America
I0267500G
500g Each for $748.63
 
Specifications

Chemical Identifiers

6439-57-2
185.199
DYYFCJRYZVHEKQ-UHFFFAOYSA-M
21225725
CCCCOC(=O)C(=C)CC(=O)[O-]
C9H13O4-
MFCD00046533
beta-Butyl Itaconate, 4-Butyl Methylenesuccinate, Itaconic Acid Monobutyl Ester
3-butoxycarbonylbut-3-enoate

Specifications

White-Yellow
6439-57-2
MFCD00046533
DYYFCJRYZVHEKQ-UHFFFAOYSA-M
3-butoxycarbonylbut-3-enoate
21225725
Crystal-Lump at 20°C
Monobutyl Itaconate
25g
C9H13O4-
beta-Butyl Itaconate, 4-Butyl Methylenesuccinate, Itaconic Acid Monobutyl Ester
CCCCOC(=O)C(=C)CC(=O)[O-]
185.199
186.21
≥95.0% (GC)
Safety and Handling

Safety and Handling

EINECSNumber : (2)-1127

TSCA : No