missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mono-tert-butyl Malonate 97.0+%, TCI America™
Supplier: TCI America M268725G
Specifications
| Mono-tert-butyl Malonate | |
| 19°C | |
| 90°C | |
| MFCD00191886 | |
| 3-tert-butoxy-3-oxopropanoic acid, malonic acid mono-tert-butyl ester, mono-tert-butyl malonate, 2-tert-butoxycarbonyl acetic acid, tert-butyl hydrogen malonate, mono-t-butyl malonate, pubchem13754, malonicacidmono-tert-butylester, malonic acid mono t-butyl ester | |
| CC(C)(C)OC(=O)CC(=O)O | |
| 160.169 | |
| 160.17 | |
| Liquid |
| 40052-13-9 | |
| Yellow | |
| C7H12O4 | |
| 25 g | |
| NGGGZUAEOKRHMA-UHFFFAOYSA-N | |
| 3-[(2-methylpropan-2-yl)oxy]-3-oxopropanoic acid | |
| 545853 | |
| ≥97.0% (GC,T) |
Chemical Identifiers
| 40052-13-9 | |
| 160.169 | |
| NGGGZUAEOKRHMA-UHFFFAOYSA-N | |
| 545853 | |
| CC(C)(C)OC(=O)CC(=O)O |
| C7H12O4 | |
| MFCD00191886 | |
| 3-tert-butoxy-3-oxopropanoic acid, malonic acid mono-tert-butyl ester, mono-tert-butyl malonate, 2-tert-butoxycarbonyl acetic acid, tert-butyl hydrogen malonate, mono-t-butyl malonate, pubchem13754, malonicacidmono-tert-butylester, malonic acid mono t-butyl ester | |
| 3-[(2-methylpropan-2-yl)oxy]-3-oxopropanoic acid |
Safety and Handling
TSCA : No