missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

mono-tert-Butyl malonate, 95%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC438830010

 View more versions of this product

Catalog No. AC438830010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

mono-tert-Butyl malonate
Glass bottle
Colorless
1g
94.0
95%
2-tert-butoxycarbonyl acetic acid, 3-tert-butoxy-3-oxopropanoic acid, malonic acid mono t-butyl ester, malonic acid mono-tert-butyl ester, malonicacidmono-tert-butylester, mono-t-butyl malonate, mono-tert-butyl malonate, pubchem13754, tert-butyl hydrogen malonate
CC(C)(C)OC(=O)CC(=O)O
160.17
160.17
95%
Authentic
90°C (2.0mmHg)
19°C to 20°C
40052-13-9
100.0
C7H12O4
NGGGZUAEOKRHMA-UHFFFAOYSA-N
3-[(2-methylpropan-2-yl)oxy]-3-oxopropanoic acid
545853
Liquid

Chemical Identifiers

40052-13-9
160.17
2-tert-butoxycarbonyl acetic acid, 3-tert-butoxy-3-oxopropanoic acid, malonic acid mono t-butyl ester, malonic acid mono-tert-butyl ester, malonicacidmono-tert-butylester, mono-t-butyl malonate, mono-tert-butyl malonate, pubchem13754, tert-butyl hydrogen malonate
3-[(2-methylpropan-2-yl)oxy]-3-oxopropanoic acid
C7H12O4
NGGGZUAEOKRHMA-UHFFFAOYSA-N
545853
CC(C)(C)OC(=O)CC(=O)O
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.
Fatal if inhaled.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Immediately call a POISON CENTER or doctor/physician.

GHS Signal Word: Danger

SDS Documents