missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
mono-tert-Butyl malonate, 95%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC438830100
Specifications
mono-tert-Butyl malonate | |
Glass bottle | |
Colorless | |
10g | |
94.0 | |
95% | |
2-tert-butoxycarbonyl acetic acid, 3-tert-butoxy-3-oxopropanoic acid, malonic acid mono t-butyl ester, malonic acid mono-tert-butyl ester, malonicacidmono-tert-butylester, mono-t-butyl malonate, mono-tert-butyl malonate, pubchem13754, tert-butyl hydrogen malonate | |
CC(C)(C)OC(=O)CC(=O)O | |
160.17 | |
160.17 | |
95% |
Authentic | |
90°C (2.0mmHg) | |
19°C to 20°C | |
40052-13-9 | |
100.0 | |
C7H12O4 | |
NGGGZUAEOKRHMA-UHFFFAOYSA-N | |
3-[(2-methylpropan-2-yl)oxy]-3-oxopropanoic acid | |
545853 | |
Liquid |
Chemical Identifiers
40052-13-9 | |
160.17 | |
2-tert-butoxycarbonyl acetic acid, 3-tert-butoxy-3-oxopropanoic acid, malonic acid mono t-butyl ester, malonic acid mono-tert-butyl ester, malonicacidmono-tert-butylester, mono-t-butyl malonate, mono-tert-butyl malonate, pubchem13754, tert-butyl hydrogen malonate | |
3-[(2-methylpropan-2-yl)oxy]-3-oxopropanoic acid |
C7H12O4 | |
NGGGZUAEOKRHMA-UHFFFAOYSA-N | |
545853 | |
CC(C)(C)OC(=O)CC(=O)O |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
Fatal if inhaled.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Immediately call a POISON CENTER or doctor/physician.
GHS Signal Word: Danger