missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

mono-Methyle5-nitroisophthalate, 98%, ACROS Organics™

Chemical Identifiers

CAS 1955-46-0
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Molecular Formula C9H7NO6
Formula Weight 225.16
MDL Number MFCD00009793
Chemical Name or Material mono-Methyle5-nitroisophthalate
Assay ≥97%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC217751000 SDS Thermo Fisher Scientific
217751000
Glass bottle 100g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

1955-46-0
100.0
C9H7NO6
MFCD00009793
≥97%
97.5
98%
225.16
mono-Methyle5-nitroisophthalate

Specifications

225.16
100g
181° to 185°C
Authentic
1955-46-0
MFCD00009793
Glass bottle
≥97%
Fine Crystalline Powder
Slightly yellow
100.0
C9H7NO6
98%
97.5
mono-Methyle5-nitroisophthalate