Please login to your online account to display your discounted pricing

mono-Methyle5-nitroisophthalate, 98%, ACROS Organics™


Quantity 100g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  217751000

 View more versions of this product

Catalog No. AC217751000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥97%
Physical Form Fine Crystalline Powder
Formula Weight 225.16
Quantity 100g
Color Slightly yellow
Melting Point 181° to 185°C
CAS Max % 100.0
Chemical Name or Material mono-Methyle5-nitroisophthalate
Infrared Spectrum Authentic
Assay Percent Range 98%
Molecular Formula C9H7NO6
MDL Number MFCD00009793
CAS 1955-46-0
Packaging Glass bottle
CAS Min % 97.5

Specifications

Chemical Name or Material mono-Methyle5-nitroisophthalate
CAS 1955-46-0
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97%
Assay Percent Range 98%
Molecular Formula C9H7NO6
Formula Weight 225.16
MDL Number MFCD00009793