Please login to your online account to display your discounted pricing

mono-Methyl Succinate 98%, ACROS Organics™


Quantity 25g
Packaging Glass bottle

Manufacturer: fisher scientific acros  127730250

 View more versions of this product

Catalog No. AC127730250

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Physical Form Crystals or Crystalline Chunks
Formula Weight 132.11
Quantity 25g
Color White to off-white
Beilstein 02, 608
TSCA TSCA
EINECS Number 223-408-2
Melting Point 55° to 59°C
CAS Max % 100.0
Chemical Name or Material mono-Methyl succinate
Infrared Spectrum Authentic
Name Note 98%
Assay Percent Range 98%
Molecular Formula C5H8O4
Linear Formula HO2CCH2CH2CO2CH3
MDL Number MFCD00002788
CAS 3878-55-5
Packaging Glass bottle
Synonym Succinic acid monomethyl ester
CAS Min % 97.5

Specifications

Chemical Name or Material mono-Methyl succinate
Name Note 98%
CAS 3878-55-5
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C5H8O4
Linear Formula HO2CCH2CH2CO2CH3
Formula Weight 132.11
MDL Number MFCD00002788
Synonym Succinic acid monomethyl ester