missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Mono-4-nitrobenzyl Malonate, TCI America™

$168.06 - $429.31

Chemical Identifiers

CAS 77359-11-6
Molecular Formula C10H9NO6
Molecular Weight (g/mol) 239.183
MDL Number MFCD00191556
InChI Key RIGFMUNSTCPGNP-UHFFFAOYSA-N
Synonym Malonic Acid Mono-4-nitrobenzyl Ester, 4-Nitrobenzyl Hydrogen Malonate
PubChem CID 592642
SMILES C1=CC(=CC=C1COC(=O)CC(=O)O)[N+](=O)[O-]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M11775G View Documents TCI America
M11775G
5g Each for $168.06
 
M117725G View Documents TCI America
M117725G
25g Each for $429.31
 
Specifications

Chemical Identifiers

77359-11-6
239.183
RIGFMUNSTCPGNP-UHFFFAOYSA-N
592642
C10H9NO6
MFCD00191556
Malonic Acid Mono-4-nitrobenzyl Ester, 4-Nitrobenzyl Hydrogen Malonate
C1=CC(=CC=C1COC(=O)CC(=O)O)[N+](=O)[O-]

Specifications

White-Yellow
5g
C10H9NO6
Malonic Acid Mono-4-nitrobenzyl Ester, 4-Nitrobenzyl Hydrogen Malonate
C1=CC(=CC=C1COC(=O)CC(=O)O)[N+](=O)[O-]
592642
Crystal-Powder at 20°C
Mono-4-nitrobenzyl Malonate
109°C
77359-11-6
MFCD00191556
RIGFMUNSTCPGNP-UHFFFAOYSA-N
239.183
239.18
≥98.0% (T)
Safety and Handling

Safety and Handling

EINECSNumber : (3)-4274

TSCA : No