missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Mono-2-O-(p-toluenesulfonyl)-gamma-cyclodextrin, TCI America™

$117.64 - $311.67

Chemical Identifiers

CAS 97227-32-2
Molecular Formula C55H86O42S
Molecular Weight (g/mol) 1451.311
InChI Key USRILNNILPQZNC-SCLKBCEUSA-N
PubChem CID 11018749
SMILES CC1=CC=C(C=C1)S(=O)(=O)OC2C(C3C(OC2OC4C(OC(C(C4O)O)OC5C(OC(C(C5O)O)OC6C(OC(C(C6O)O)OC7C(OC(C(C7O)O)OC8C(OC(C(C8O)O)OC9C(OC(C(C9O)O)OC1C(OC(O3)C(C1O)O)CO)CO)CO)CO)CO)CO)CO)CO)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M19571G View Documents TCI America
M19571G
1g Each for $311.67
 
M1957200MG View Documents TCI America
M1957200MG
200mg Each for $117.64
 
Specifications

Chemical Identifiers

97227-32-2
1451.311
11018749
C55H86O42S
USRILNNILPQZNC-SCLKBCEUSA-N
CC1=CC=C(C=C1)S(=O)(=O)OC2C(C3C(OC2OC4C(OC(C(C4O)O)OC5C(OC(C(C5O)O)OC6C(OC(C(C6O)O)OC7C(OC(C(C7O)O)OC8C(OC(C(C8O)O)OC9C(OC(C(C9O)O)OC1C(OC(O3)C(C1O)O)CO)CO)CO)CO)CO)CO)CO)CO)O

Specifications

White-Yellow
97227-32-2
USRILNNILPQZNC-SCLKBCEUSA-N
1451.311
1451.31
≥95.0% (HPLC)
1g
C55H86O42S
CC1=CC=C(C=C1)S(=O)(=O)OC2C(C3C(OC2OC4C(OC(C(C4O)O)OC5C(OC(C(C5O)O)OC6C(OC(C(C6O)O)OC7C(OC(C(C7O)O)OC8C(OC(C(C8O)O)OC9C(OC(C(C9O)O)OC1C(OC(O3)C(C1O)O)CO)CO)CO)CO)CO)CO)CO)CO)O
11018749
Crystal-Powder at 20°C
Mono-2-O-(p-toluenesulfonyl)-gamma-cyclodextrin
Safety and Handling

Safety and Handling

TSCA : No