missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Mono-2-O-(p-toluenesulfonyl)-beta-cyclodextrin Hydrate, TCI America™

$134.45 - $362.09

Chemical Identifiers

CAS 84216-71-7
Molecular Formula C49H76O37S
Molecular Weight (g/mol) 1289.17
InChI Key VGZWDYWXWGKKEE-UJPGXMRNSA-N
PubChem CID 11829278
SMILES CC1=CC=C(C=C1)S(=O)(=O)OC2C(C3C(OC2OC4C(OC(C(C4O)O)OC5C(OC(C(C5O)O)OC6C(OC(C(C6O)O)OC7C(OC(C(C7O)O)OC8C(OC(C(C8O)O)OC9C(OC(O3)C(C9O)O)CO)CO)CO)CO)CO)CO)CO)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M17411G View Documents TCI America
M17411G
1g Each for $362.09
 
M1741200MG View Documents TCI America
M1741200MG
200mg Each for $134.45
 
Specifications

Chemical Identifiers

84216-71-7
1289.17
11829278
C49H76O37S
VGZWDYWXWGKKEE-UJPGXMRNSA-N
CC1=CC=C(C=C1)S(=O)(=O)OC2C(C3C(OC2OC4C(OC(C(C4O)O)OC5C(OC(C(C5O)O)OC6C(OC(C(C6O)O)OC7C(OC(C(C7O)O)OC8C(OC(C(C8O)O)OC9C(OC(O3)C(C9O)O)CO)CO)CO)CO)CO)CO)CO)O

Specifications

White
84216-71-7
VGZWDYWXWGKKEE-UJPGXMRNSA-N
1289.17
1289.17
≥97.0% (HPLC)
1g
C49H76O37S
CC1=CC=C(C=C1)S(=O)(=O)OC2C(C3C(OC2OC4C(OC(C(C4O)O)OC5C(OC(C(C5O)O)OC6C(OC(C(C6O)O)OC7C(OC(C(C7O)O)OC8C(OC(C(C8O)O)OC9C(OC(O3)C(C9O)O)CO)CO)CO)CO)CO)CO)CO)O
11829278
Crystal-Powder at 20°C
Mono-2-O-(p-toluenesulfonyl)-beta-cyclodextrin Hydrate
Safety and Handling

Safety and Handling

TSCA : No