missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Mono-2-O-(p-toluenesulfonyl)-alpha-cyclodextrin, TCI America™

$110.00 - $311.67

Chemical Identifiers

CAS 93184-10-2
Molecular Formula C43H66O32S
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M1956200MG View Documents TCI America
M1956200MG
200mg Each for $110.00
 
M19561G View Documents TCI America
M19561G
1g Each for $311.67
 
Specifications

Chemical Identifiers

93184-10-2
C43H66O32S

Specifications

White-Yellow
200mg
C43H66O32S
Crystal-Powder at 20°C
Mono-2-O-(p-toluenesulfonyl)-alpha-cyclodextrin
176°C
93184-10-2
1127.03
≥98.0% (HPLC)
Safety and Handling

Safety and Handling

TSCA : No