missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Mono-2-O-(p-toluenesulfonyl)-alpha-cyclodextrin 98.0+%, TCI America™
Supplier: TCI America M19561G
Specifications
Mono-2-O-(p-toluenesulfonyl)-alpha-cyclodextrin | |
176°C | |
C43H66O32S | |
1127.03 | |
Crystalline Powder |
93184-10-2 | |
White-Yellow | |
1 g | |
≥98.0% (HPLC) |
Chemical Identifiers
93184-10-2 | |
C43H66O32S |
Safety and Handling
TSCA : No