Please login to your online account to display your discounted pricing

eBioscience™ Monkey sTNF-R I Platinum ELISA Kit

For the quantitative detection of monkey sTNF-R I.

$520.84 - $2,932.34

Specifications

For Use With (Application) ELISA
Sample Size 10μL
Synonym CD120a, TNFR60, TNF Receptor superfamily member 1A
Target sCD120a (sTNF Receptor I)
Conjugate HRP
View More Specs
Includes:  Aluminium pouch(es) with a Microwell Plate coated with monoclonal antibody to monkey sTNF-R I
HRP-Conjugate anti-monkey sTNF-R I monoclonal antibody
Monkey sTNF-R I Standard concentrate, 12 U/ml upon dilution
Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA)
Wash Buffer Concentrate 20x (PBS with 1% Tween 20)
Substrate Solution (tetramethyl-benzidine)
Stop Solution (1M Phosphoric acid)
Blue-Dye
Green-Dye
Adhesive Films

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501125476

 
life technologies
LSBMS653TEN
10 x 96 tests Each for $2,932.34

5018403

 
life technologies
LSBMS653
96 tests Each for $520.84
Description & Specifications

Specifications

For Use With (Application) ELISA
Sample Size 10μL
Synonym CD120a, TNFR60, TNF Receptor superfamily member 1A
Target sCD120a (sTNF Receptor I)
Conjugate HRP
Sample Type Cell culture supernatant, serum (baboon, cynomolgus)
Sensitivity 0.06 U/mL
Species Monkey

Platinum Kits undergo stringent in-house development, manufacture and quality-validation processes ensuring highest possible product performance and optimal lot-to-lot consistency