missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Invitrogen™ eBioscience™ Monkey sTNF-R I Platinum ELISA Kit

For the quantitative detection of monkey sTNF-R I.

Manufacturer:  Invitrogen™ LSBMS653TEN

 View more versions of this product

Catalog No. 501125476



Includes: Aluminium pouch(es) with a Microwell Plate coated with monoclonal antibody to monkey sTNF-R I
HRP-Conjugate anti-monkey sTNF-R I monoclonal antibody
Monkey sTNF-R I Standard concentrate, 12 U/ml upon dilution
Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA)
Wash Buffer Concentrate 20x (PBS with 1% Tween 20)
Substrate Solution (tetramethyl-benzidine)
Stop Solution (1M Phosphoric acid)
Blue-Dye
Green-Dye
Adhesive Films

Description

Description

Platinum Kits undergo stringent in-house development, manufacture and quality-validation processes ensuring highest possible product performance and optimal lot-to-lot consistency
Specifications

Specifications

HRP
10 x 96 tests
Cell culture supernatant, serum (baboon, cynomolgus)
Monkey
CD120a, TNFR60, TNF Receptor superfamily member 1A
ELISA
0.06 U/mL
10μL
sCD120a (sTNF Receptor I)
Documents

For Research Use Only