Please login to your online account to display your discounted pricing

eBioscience Monkey sPECAM-1 Platinum ELISA Kit

For the quantitative detection of soluble monkey platelet endothelial cell adhesion molecule-1.

$651.05 - $4,189.05

Specifications

For Use With (Application) ELISA
Sample Size 10μL
Synonym Leukocyte surface protein, CD31, EndoCAM, HEC7
Target sCD31 (sPECAM-1)
Conjugate HRP
View More Specs
Includes:  Aluminium pouch(es) with a Microwell Plate coated with Monoclonal Antibody (murine) to monkey sPECAM-1
HRP-Conjugate anti-monkey-sPECAM-1 monoclonal (murine) antibody
Monkey sPECAM-1 Standard
Wash Buffer Concentrate 20x (PBS with 1% Tween 20)
Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA)
Sample Diluent (buffered protein matrix)
Substrate Solution (tetramethyl-benzidine)
Stop Solution (1M Phosphoric acid)
Blue-Dye
Green-Dye
Adhesive Films

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5018396

 
ebioscience
LSBMS651
96 tests Each for $651.05

5018399

 
ebioscience
LSBMS651TEN
10 x 96 tests Each for $4,189.05
Description & Specifications

Specifications

For Use With (Application) ELISA
Sample Size 10μL
Synonym Leukocyte surface protein, CD31, EndoCAM, HEC7
Target sCD31 (sPECAM-1)
Conjugate HRP
Sample Type Cell culture supernatant, serum (baboon)
Sensitivity 0.06 U/mL
Species Monkey

Platinum Kits undergo stringent in-house development, manufacture and quality-validation processes ensuring highest possible product performance and optimal lot-to-lot consistency