Please login to your online account to display your discounted pricing

eBioscience monkey sP-selectin Platinum ELISA Kit

For the quantitative detection of monkey sP-selectin.

$622.05 - $4,189.05

Specifications

For Use With (Application) ELISA
Sample Size 10μL
Synonym Granule membrane protein 140, PADGEM, CD62P
Target sCD62P (sP-Selectin)
Conjugate HRP
View More Specs
Includes:  Aluminium pouch(es) with a Microwell Plate coated with monoclonal antibody to monkey sP-selectin
HRP-Conjugate anti-monkey sP-selectin monoclonal antibody
Monkey sP-selectin Standard lyophilized, 100.0 U/ml upon reconstitution
Sample Diluent
Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA)
Wash Buffer Concentrate 20x (PBS with 1% Tween 20)
Substrate Solution (tetramethyl-benzidine)
Stop Solution (1M Phosphoric acid)
Blue-Dye
Green-Dye
Adhesive Films

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5018394

 
ebioscience
BMS6504
96 tests Each for $622.05

5018395

 
ebioscience
BMS6504TEN
10 x 96 tests Each for $4,189.05
Description & Specifications

Specifications

For Use With (Application) ELISA
Sample Size 10μL
Synonym Granule membrane protein 140, PADGEM, CD62P
Target sCD62P (sP-Selectin)
Conjugate HRP
Sample Type Cell culture supernatant, serum (baboon)
Sensitivity 1.3 U/mL
Species Monkey

Platinum Kits undergo stringent in-house development, manufacture and quality-validation processes ensuring highest possible product performance and optimal lot-to-lot consistency