Please login to your online account to display your discounted pricing

eBioscience™ Monkey sE-selectin Platinum ELISA Kit

For the quantitative detection of monkey sE-selectin.

$520.84 - $2,932.34

Specifications

For Use With (Application) ELISA
Sample Size 20μL
Synonym ELAM1, endothelial adhesion molecule 1, CD62E
Target sCD62E (sE-Selectin)
Conjugate HRP
View More Specs
Includes:  Aluminium pouch(es) with a Microwell Plate coated with monoclonal antibody to monkey sE-selectin
HRP-Conjugate anti-monkey sE-selectin monoclonal antibody
Monkey sE-selectin Standard lyophilized, 120 U/ml upon reconstitution
Sample Diluent
Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA)
Wash Buffer Concentrate 20x (PBS with 1% Tween 20)
Substrate Solution (tetramethyl-benzidine)
Stop Solution (1M Phosphoric acid)
Blue-Dye
Green-Dye
Adhesive Films

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5018391

 
life technologies
LSBMS649
96 tests Each for $520.84

5018393

 
life technologies
LSBMS649TEN
10 x 96 tests Each for $2,932.34
Description & Specifications

Specifications

For Use With (Application) ELISA
Sample Size 20μL
Synonym ELAM1, endothelial adhesion molecule 1, CD62E
Target sCD62E (sE-Selectin)
Conjugate HRP
Sample Type Cell culture supernatant, serum (baboon)
Sensitivity 0.4 U/mL
Species Monkey

Platinum Kits undergo stringent in-house development, manufacture and quality-validation processes ensuring highest possible product performance and optimal lot-to-lot consistency