Please login to your online account to display your discounted pricing

eBioscience™ Monkey IL-8 Platinum ELISA Kit

For the quantitative detection of monkey IL-8.

$651.05 - $4,189.05

Specifications

For Use With (Application) ELISA
Sample Size 50μL
Synonym Protein 3-10C, CXCL8, NAP-1, GCP-1
Target IL-8
Conjugate Biotin
View More Specs
Includes:  Aluminium pouch(es) with a Microwell Plate coated with monoclonal antibody to monkey IL-8
Biotin-Conjugate anti-monkey IL-8 polyclonal antibody
Streptavidin-HRP
Monkey IL-8 Standard Concentrate, 125000 U/ml
Sample Diluent
Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA)
Wash Buffer Concentrate 20x (PBS with 1% Tween 20)
Substrate Solution (tetramethyl-benzidine)
Stop Solution (1M Phosphoric acid)
Blue-Dye
Green-Dye
Red-Dye
Adhesive Films

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5018370

 
life technologies
LSBMS6403
96 tests Each for $651.05

5018373

 
life technologies
LSBMS6403TEN
10 x 96 tests Each for $4,189.05
Description & Specifications

Specifications

For Use With (Application) ELISA
Sample Size 50μL
Synonym Protein 3-10C, CXCL8, NAP-1, GCP-1
Target IL-8
Conjugate Biotin
Sample Type Cell culture supernatant, serum (baboon, cynomolgus)
Sensitivity 3.0 U/mL
Species Monkey

Platinum Kits undergo stringent in-house development, manufacture and quality-validation processes ensuring highest possible product performance and optimal lot-to-lot consistency