missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Invitrogen™ eBioscience™ Monkey IL-2 Platinum ELISA Kit
For the quantitative detection of monkey IL-2.
Manufacturer: Invitrogen™ LSBMS6432
Includes: Aluminium pouch(es) with a Microwell Plate coated with monoclonal antibody to monkey IL-2
Description
Interleukin 2 (IL-2) is an immuno-modulatory cytokine that is important for the proliferation of activated T cells, differentiation of B cells, natural killer cells, monocytes and macrophages. IL-2 signals through the IL-2 receptor (IL-2R), a heterotrimeric protein complex whose gamma chain is also shared by interleukin 4 (IL-4) and interleukin 7 (IL-7). The expression of the IL-2 gene in mature thymocytes is monoallelic, which represents an unusual regulatory mode for controlling the precise expression of a single gene. The targeted disruption of a gene similar to IL-2 in mice leads to an ulcerative colitis-like disease that suggests an essential role of this gene in the immune response to antigenic stimuli.Specifications
Q29615, Q865Y1 | |
Biotin | |
100126754, 102129830 | |
Pre-coated 96 well plate, Standard, Sample Diluent, Assay Buffer concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers | |
96 tests | |
9.1 U/mL | |
50μL | |
Monkey | |
1 hr 20 min | |
Baboon, Cynomolgus Monkey | |
Aldesleukin, T cell growth factor, TCGF | |
7% |
18.8-1,200 U/mL | |
ELISA | |
Colorimetric Microplate Reader | |
ELISA | |
RUO | |
Amniotic Fluid, Plasma, Serum, Supernatants, Urine, Whole Blood | |
Amniotic Fluid (50μL), Plasma (50μL), Serum (50μL), Supernatants (50μL), Urine (50μL), Whole Blood (50μL) | |
2-8°C | |
3 hr 30 min | |
IL2 | |
5% | |
HRP |