Invitrogen™ eBioscience™ Monkey IL-2 Platinum ELISA Kit
For the quantitative detection of monkey IL-2.
Manufacturer:  Invitrogen™ LSBMS6432FIVE
 View more versions of this product
Catalog No. 501125473
Includes: Aluminium pouch(es) with a Microwell Plate coated with monoclonal antibody to monkey IL-2
Description
Description
Platinum Kits undergo stringent in-house development, manufacture and quality-validation processes ensuring highest possible product performance and optimal lot-to-lot consistency
Specifications
Specifications
ELISA
9.1 U/mL
50μL
IL-2
5 x 96 tests
Cell culture supernatant, serum, plasma (EDTA, citrate, heparin)
Monkey
Aldesleukin, T cell growth factor, TCGF
Documents