Please login to your online account to display your discounted pricing

eBioscience Monkey IL-12 (p70) Platinum ELISA Kit

For the quantitative detection of monkey IL-12 p70.

$651.05 - $4,189.05

Specifications

For Use With (Application) ELISA
Sample Size 50μL
Synonym Interleukin-12 subunit p70, CLMF, NKSF
Target IL-12
Conjugate Biotin
View More Specs
Includes:  Aluminium pouch(es) with a Microwell Plate coated with monoclonal antibody to monkey IL-12 p70
Biotin-Conjugate anti-monkey IL-12 p70 monoclonal antibody
Streptavidin-HRP
Monkey IL-12 p70 Standard lyophilized, 500 U/ml upon reconstitution
Sample Diluent
Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA)
Wash Buffer Concentrate 20x (PBS with 1% Tween 20)
Substrate Solution (tetramethyl-benzidine)
Stop Solution (1M Phosphoric acid)
Blue-Dye
Green-Dye
Red-Dye
Adhesive Films

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5018384

 
ebioscience
LSBMS646
96 tests Each for $651.05

5018386

 
ebioscience
LSBMS646TEN
10 x 96 tests Each for $4,189.05
Description & Specifications

Specifications

For Use With (Application) ELISA
Sample Size 50μL
Synonym Interleukin-12 subunit p70, CLMF, NKSF
Target IL-12
Conjugate Biotin
Sample Type Cell culture supernatant, serum (baboon)
Sensitivity 2.6 U/mL
Species Monkey

Platinum Kits undergo stringent in-house development, manufacture and quality-validation processes ensuring highest possible product performance and optimal lot-to-lot consistency