missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
GIBCO™ Monkey (cynomolgus) Microsomes
Liver microsomes prepared from a pool of male Cynomologous Monkeys. Total protein concentration: 20mg/mL.
Supplier: GIBCO™ MKMCPL
Specifications
Male | |
Pooled | |
Research Grade | |
20 mg/mL | |
Store between -60°C and -85°C. | |
Microsomes | |
Gibco™ |
Frozen | |
Liver | |
Monkey | |
Monkey Microsome | |
Tube | |
Dry Ice |