Please login to your online account to display your discounted pricing

Monensin, Sodium Salt 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Monensin, sodium salt
Name Note 97%
CAS 22373-78-0
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Molecular Formula C36H61NaO11
Formula Weight 692.85
MDL Number MFCD00077826
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC415110250

  SDS
thermo fisher scientific
AC415110250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 692.85
Quantity 25g
RTECS Number JH2830000
EINECS Number 244-941-7
Specific Rotation Condition +57.30° (20°C c=4.0,in methanol)
Molecular Formula C36H61NaO11
Packaging Glass bottle
CAS Min % 96.0
Health Hazard 1 GHS Signal Word: Danger
CAS Max % 100.0
Health Hazard 2 GHS H Statement:
Toxic if swallowed.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Chemical Name or Material Monensin, sodium salt
Name Note 97%
Assay Percent Range 97%
MDL Number MFCD00077826
CAS 22373-78-0