Please login to your online account to display your discounted pricing

Monensin, Sodium Salt 97%, ACROS Organics™


Quantity 25g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC415110250

 View more versions of this product

Catalog No. AC415110250

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 692.85
Quantity 25g
RTECS Number JH2830000
EINECS Number 244-941-7
Specific Rotation Condition +57.30° (20°C c=4.0,in methanol)
Health Hazard 1 GHS Signal Word: Danger
CAS Min % 96.0
CAS Max % 100.0
Packaging Glass bottle
Health Hazard 2 GHS H Statement:
Toxic if swallowed.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Molecular Formula C36H61NaO11
CAS 22373-78-0
MDL Number MFCD00077826
Name Note 97%
Chemical Name or Material Monensin, sodium salt
Assay Percent Range 97%

Specifications

Chemical Name or Material Monensin, sodium salt
Name Note 97%
CAS 22373-78-0
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Molecular Formula C36H61NaO11
Formula Weight 692.85
MDL Number MFCD00077826
Accessories

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Toxic if swallowed.

SDS & Safety Documents