missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Monensin sodium salt, 50mg/mL in methanol, sterile-filtered, Alfa Aesar™

Manufacturer:  Alfa Aesar J67431XF

 View more versions of this product

Catalog No. AAJ67431XF



Specifications

Specifications

Monensin sodium salt
C36H61NaO11
UN1230
XOIQMTLWECTKJL-JXDONITHSA-M
sodium;(3S,4R)-4-[(2R,5S,7R,8S,9R)-2-[(2S,5R)-5-ethyl-5-[(2S,3R,5S)-5-[(2R,3R,5S,6S)-6-hydroxy-6-(hydroxymethyl)-3,5-dimethyloxan-2-yl]-3-methyloxolan-2-yl]oxolan-2-yl]-7-hydroxy-2,8-dimethyl-1,10-dioxaspiro[4.5]decan-9-yl]-3-methoxy-2-methylpentanoate
133612076
1mL
22373-78-0
MFCD00077826
Plastic bottle
CCC1(CCC(O1)C2(CCC3(O2)CC(C(C(O3)C(C)C(C(C)C(=O)[O-])OC)C)O)C)C4C(CC(O4)C5C(CC(C(O5)(CO)O)C)C)C.[Na+]
692.863
Liquid

Chemical Identifiers

22373-78-0
692.863
XOIQMTLWECTKJL-JXDONITHSA-M
sodium;(3S,4R)-4-[(2R,5S,7R,8S,9R)-2-[(2S,5R)-5-ethyl-5-[(2S,3R,5S)-5-[(2R,3R,5S,6S)-6-hydroxy-6-(hydroxymethyl)-3,5-dimethyloxan-2-yl]-3-methyloxolan-2-yl]oxolan-2-yl]-7-hydroxy-2,8-dimethyl-1,10-dioxaspiro[4.5]decan-9-yl]-3-methoxy-2-methylpentanoate
C36H61NaO11
MFCD00077826
133612076
CCC1(CCC(O1)C2(CCC3(O2)CC(C(C(O3)C(C)C(C(C)C(=O)[O-])OC)C)O)C)C4C(CC(O4)C5C(CC(C(O5)(CO)O)C)C)C.[Na+]
Safety and Handling

Safety and Handling

TSCA : No

Documents