Please login to your online account to display your discounted pricing

Monastrol, Tocris


Selective inhibitor of mitotic kinesin Eg5

Quantity 10mg

Manufacturer: r & d systems  130510

 View more versions of this product

Catalog No. 130510

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 292.35
Quantity 10mg
Chemical Name or Material Monastrol
Assay Percent Range >98%
CAS 254753-54-3
Synonym 1,2,3,4-Tetrahydro-4-(3-hydroxyphenyl)-6-methyl-2-thioxo-5-pyrimidinecarboxylic acid, ethyl ester
Molecular Formula C14H16N2O3S
Recommended Storage Store at Room Temperature

Specifications

Chemical Name or Material Monastrol
Assay Percent Range >98%
Molecular Formula C14H16N2O3S
Formula Weight 292.35
Synonym 1,2,3,4-Tetrahydro-4-(3-hydroxyphenyl)-6-methyl-2-thioxo-5-pyrimidinecarboxylic acid, ethyl ester
Accessories

Store at Room Temperature