Please login to your online account to display your discounted pricing

Alfa Aesar™ Monastrol, 98%

Chemical Identifiers

Chemical Name or Material Monastrol
CAS 329689-23-8
Formula Weight 292.36
MDL Number MFCD00813077
Merck Index 14,6244
Synonym Ethyl 4-(3-hydroxyphenyl)-6-methyl-2-thioxo-1,2,3,4-tetrahydro-4H-pyrimidin-5-carboxylate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ67096LB0

  SDS
alfa aesar™
J67096LB0
5mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 292.36
Melting Point 179° to 180°C
Chemical Name or Material Monastrol
CAS 329689-23-8
MDL Number MFCD00813077
Synonym Ethyl 4-(3-hydroxyphenyl)-6-methyl-2-thioxo-1,2,3,4-tetrahydro-4H-pyrimidin-5-carboxylate
Physical Form Solid
Assay 98%
Quantity 5mg
Color White to pale yellow
Storage Note 1 -30° to -10°C
RTECS Number UV6262400
Merck Index 14,6244
Sensitivity Light Sensitive
Safety and Handling

-30° to -10°C