Molybdic Acid Dihydrate Sodium Salt, (Crystalline),MP Biomedicals
Manufacturer:  Mp Biomedicals 15170590
 View more versions of this product
Catalog No. ICN15170590
Specifications Description & Specifications
Specifications
241.9
Crystalline
Na2MoO4 · 2H2O
500g
Molybdic Acid Dihydrate Sodium Salt
10102-40-6
Chemical Identifiers
10102-40-6
Na2MoO4 · 2H2O
Molybdic Acid Dihydrate Sodium Salt
Crystalline
241.9
Safety and Handling
Recommended Storage : Room Temperature