Please login to your online account to display your discounted pricing

Molybdic Acid Dihydrate Sodium Salt, (Crystalline),MP Biomedicals

$67.97 - $227.12

Chemical Identifiers

Chemical Name or Material Molybdic Acid Dihydrate Sodium Salt
CAS 10102-40-6
Physical Form Crystalline
Molecular Formula Na2MoO4 · 2H2O
Formula Weight 241.9
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

ICN15170590

 
mp biomedicals
15170590
500g Each for $227.12

ICN15170580

 
mp biomedicals
15170580
100g Each for $67.97
Description & Specifications

Specifications

Formula Weight 241.9
Quantity 500g
Physical Form Crystalline
Chemical Name or Material Molybdic Acid Dihydrate Sodium Salt
Molecular Formula Na2MoO4 · 2H2O
CAS 10102-40-6
Recommended Storage Room Temperature
Safety and Handling

Room Temperature