Learn More
Molybdic acid, ACS, MoO{3} 85% min, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 02298336
Description
Molybdic acid is used as a precursor in the production of ammonium molybdate tetrahydrate and molybdenum(IV) sulfide. The mixture of molybdic acid in concentrated sulfuric acid is called as Froehde reagent, which finds application in analytical chemistry for the detection of alkaloids.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Molybdic acid | |
Crystalline Powder | |
H2MoO4 | |
molybdic acid, molybdic vi acid, unii-i96n991j1n, dihydroxy dioxo molybdenum, molybdic acid acs, dihydroxy-dioxomolybdenum, molybdate moo42-, dihydrogen, t-4, dihydroxidodioxidomolybdenum, aronis24112, moo2 oh 2 | |
VLAPMBHFAWRUQP-UHFFFAOYSA-L | |
dihydroxy(dioxo)molybdenum | |
82208 | |
ACS | |
Odorless |
7782-91-4 | |
500 g | |
MFCD00036287 | |
Soluble in dilute ammonium hydroxide,alkali and ammonia. Slightly soluble in water. | |
O[Mo](O)(=O)=O | |
161.96 | |
CHEBI:25371 | |
MoO3 85% min. | |
3.1g/mL |
Chemical Identifiers
7782-91-4 | |
161.96 | |
VLAPMBHFAWRUQP-UHFFFAOYSA-L | |
82208 | |
dihydroxy(dioxo)molybdenum |
H2MoO4 | |
MFCD00036287 | |
molybdic acid, molybdic vi acid, unii-i96n991j1n, dihydroxy dioxo molybdenum, molybdic acid acs, dihydroxy-dioxomolybdenum, molybdate moo42-, dihydrogen, t-4, dihydroxidodioxidomolybdenum, aronis24112, moo2 oh 2 | |
CHEBI:25371 | |
O[Mo](O)(=O)=O |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P260-P264b-P271-P280i-P304+P340-P305+P351+P338-P312-P314-P501c
H319-H335-H373
EINECSNumber : 231-970-5
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only