missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Molybdenum wire, 2mm diameter, 99.95% (metals basis), Alfa Aesar™

Manufacturer:  Thermo Fisher Scientific 10040G1

 View more versions of this product

Catalog No. AA10040G1



Specifications

Specifications

Molybdenum
7439-98-7
99.95% (Metals basis)
MFCD00003465
ZOKXTWBITQBERF-UHFFFAOYSA-N
molybdenum
95.95
CHEBI:28685
1m
Dark gray wire
Mo
6+ ion, amperit 106.2, atom, mchvl, metco 63, molybdenum, elemental, molybdenum, ion 6+, molybdenum, metallic, tsm1, vi ion
[Mo]
Odorless
23932
Wire, ≈32.1g/m,Temper: soft

Chemical Identifiers

7439-98-7
95.95
ZOKXTWBITQBERF-UHFFFAOYSA-N
23932
molybdenum
Mo
MFCD00003465
6+ ion, amperit 106.2, atom, mchvl, metco 63, molybdenum, elemental, molybdenum, ion 6+, molybdenum, metallic, tsm1, vi ion
CHEBI:28685
[Mo]
Safety and Handling

Safety and Handling

EINECSNumber : 231-107-2

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents