missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Molybdenum wire, 0.25mm (0.01 in.) dia., annealed, 99.95% (metals basis)

Manufacturer:  Alfa Aesar™ 10036G9

 View more versions of this product

Catalog No. AA10036G9



Specifications

Specifications

Molybdenum wire
25m
7439-98-7
Mo
molybdenum, elemental, 6+ ion, mchvl, vi ion, molybdenum, metallic, metco 63, atom, molybdenum, ion 6+, tsm1, amperit 106.2
[Mo]
95.95
CHEBI:28685
99.95%
≈0.50g/m
0.25mm (0.01 in.) dia., Annealed, Temper: soft
(metals basis)
MFCD00003465
ZOKXTWBITQBERF-UHFFFAOYSA-N
molybdenum
23932
Wire

Chemical Identifiers

7439-98-7
95.95
ZOKXTWBITQBERF-UHFFFAOYSA-N
23932
molybdenum
Mo
MFCD00003465
molybdenum, elemental, 6+ ion, mchvl, vi ion, molybdenum, metallic, metco 63, atom, molybdenum, ion 6+, tsm1, amperit 106.2
CHEBI:28685
[Mo]
Safety and Handling

Safety and Handling

TSCA : TSCA

Documents