missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Molybdenum wire, 0.203mm (0.008in) dia, hard, 99.9%(metals basis), Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 045134G1

 View more versions of this product

Catalog No. AA45134G1



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Molybdenum wire
7439-98-7
Mo
1 m
ZOKXTWBITQBERF-UHFFFAOYSA-N
molybdenum
23932
99.9%
Wire
0.203mm (0.008 in.) dia., Hard
(metals basis)
MFCD00003465
molybdenum, elemental, 6+ ion, mchvl, vi ion, molybdenum, metallic, metco 63, atom, molybdenum, ion 6+, tsm1, amperit 106.2
[Mo]
95.95
CHEBI:28685
≈0.33g/m

Chemical Identifiers

7439-98-7
95.95
ZOKXTWBITQBERF-UHFFFAOYSA-N
23932
molybdenum
Mo
MFCD00003465
molybdenum, elemental, 6+ ion, mchvl, vi ion, molybdenum, metallic, metco 63, atom, molybdenum, ion 6+, tsm1, amperit 106.2
CHEBI:28685
[Mo]
Safety and Handling

Safety and Handling

EINECSNumber : 231-107-2

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only