Please login to your online account to display your discounted pricing

Alfa Aesar™ Molybdenum(VI) sulfide dihydrate

Chemical Identifiers

Chemical Name or Material Molybdenum(VI) sulfide dihydrate
CAS 12033-29-3
Formula Weight 228.13 (192.13 Anhydrous)
MDL Number MFCD00014226
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA3274106

  SDS
alfa aesar™
3274106
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA3274114

  SDS
alfa aesar™
3274114
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 228.13 (192.13 Anhydrous)
TSCA TSCA
CAS 12033-29-3
Physical Form Powder
Odor Odorless
Quantity 5g
Chemical Name or Material Molybdenum(VI) sulfide dihydrate
MDL Number MFCD00014226