Learn More
Molybdenum(VI) oxide, Puratronic™, 99.998% (metals basis excluding W), W 300ppm max, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 01081214
Description
Its chief application is as an oxidation catalyst and as a raw material for the production of molybdenum metal. It is a Precursor to LAMOX fast ion conductors and superconductors. Molybdenum trioxide has also been suggested as a potential anti-microbial agent, e.g., in polymers. In contact with water, it forms H+ ions that can kill bacteria effectively.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
1155°C (sublimation) | |
795°C | |
White to Gray | |
25 g | |
MoO3 | |
UN3288 | |
molybdenum trioxide, molybdenum vi oxide, molybdic anhydride, molybdic trioxide, molybdena, molybdic oxide, molybdenum oxide, molybdic acid anhydride, molybdenum peroxide | |
JKQOBWVOAYFWKG-UHFFFAOYSA-N | |
trioxomolybdenum | |
14802 | |
143.94 | |
Puratronic™ | |
4.692 g/mL |
Molybdenum(VI) oxide | |
1313-27-5 | |
Powder | |
(metals basis excluding W), W 300ppm max. | |
MFCD00003469 | |
14,6239 | |
Soluble at 0.490g/1000 cc water @ 28°C. Soluble in concentrated mineral acids. | |
O=[Mo](=O)=O | |
143.95 | |
CHEBI:30627 | |
99.998% | |
Odorless |
Chemical Identifiers
1313-27-5 | |
143.95 | |
JKQOBWVOAYFWKG-UHFFFAOYSA-N | |
14802 | |
trioxomolybdenum |
MoO3 | |
MFCD00003469 | |
molybdenum trioxide, molybdenum vi oxide, molybdic anhydride, molybdic trioxide, molybdena, molybdic oxide, molybdenum oxide, molybdic acid anhydride, molybdenum peroxide | |
CHEBI:30627 | |
O=[Mo](=O)=O |
Safety and Handling
GHS H Statement
H300-H351-H319-H335
Fatal if swallowed.
Suspected of causing cancer.
Causes serious eye irritation.
May cause respiratory irritation.
P201-P202-P261-P264b-P271-P280i-P281-P304+P340-P305+P351+P338-P308+P313-P501c
H319-H335-H351
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLID, INORGANIC, N.O.S.
EINECSNumber : 215-204-7
RTECSNumber : QA4725000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only