Please login to your online account to display your discounted pricing

Alfa Aesar™ Molybdenum(VI) oxide bis(2,4-pentanedionate), 99%

$26.16 - $48.64

Chemical Identifiers

Chemical Name or Material Molybdenum (VI) oxide bis (2,4-pentanedionate)
CAS 17524-05-9
Molecular Formula C10H14MoO6
Formula Weight 326.16
MDL Number MFCD00011506
Synonym Bis(acetylacetonato)dioxomolybdenum(VI); Molybdenyl acetylacetonate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4414009

  SDS
alfa aesar™
4414009
10g Each for $48.64

AA4414004

  SDS
alfa aesar™
4414004
2g Each for $26.16
Description & Specifications

Specifications

Formula Weight 326.16
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Chemical Name or Material Molybdenum (VI) oxide bis (2,4-pentanedionate)
Physical Form Powder
Assay 99%
Sensitivity Air Sensitive
Molecular Formula C10H14MoO6
MDL Number MFCD00011506
CAS 17524-05-9
Synonym Bis(acetylacetonato)dioxomolybdenum(VI); Molybdenyl acetylacetonate
Color Pale yellow
Melting Point ≈185°C (dec.)
Quantity 10g
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.