missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Molybdenum(VI) oxide, 99.9995% (metals basis excluding W), Alfa Aesar™, Puratronic™

Manufacturer:  Alfa Aesar 1293014

 View more versions of this product

Catalog No. AA1293014



Specifications

Specifications

Molybdenum(VI) oxide
99.9995% (metals basis excluding W)
MFCD00003469
molybdenum trioxide, molybdenum vi oxide, molybdic anhydride, molybdic trioxide, molybdena, molybdic oxide, molybdenum oxide, molybdic acid anhydride, molybdenum peroxide, molybdenum vi oxide
O=[Mo](=O)=O
143.947
CHEBI:30627
Powder
Odorless
25g
1313-27-5
MoO3
White to gray powder
JKQOBWVOAYFWKG-UHFFFAOYSA-N
trioxomolybdenum
14802
143.94
4.692g/cm3
White to Gray

Chemical Identifiers

1313-27-5
143.947
JKQOBWVOAYFWKG-UHFFFAOYSA-N
14802
trioxomolybdenum
MoO3
MFCD00003469
molybdenum trioxide, molybdenum vi oxide, molybdic anhydride, molybdic trioxide, molybdena, molybdic oxide, molybdenum oxide, molybdic acid anhydride, molybdenum peroxide, molybdenum vi oxide
CHEBI:30627
O=[Mo](=O)=O
Safety and Handling

Safety and Handling

H300-H319-H335-H351: Fatal if swallowed. Causes serious eye irritation. May cause respiratory irritation. Suspected of causing cancer.

DOTInformation : Hazard Class: 6.1, Packing Group: III

EINECSNumber : 215-204-7

Documents