Please login to your online account to display your discounted pricing

Molybdenum(VI) oxide, 99.999%, (trace metal basis), ACROS Organics™

Chemical Identifiers

Chemical Name or Material Molybdenum(VI) oxide
CAS 1313-27-5
CAS Min % 100.0
CAS Max % 100.0
Purity Grade Trace Metal Basis
Assay 64.3 to 69.0%
Assay Percent Range 99.999%
Molecular Formula MoO3
Linear Formula MoO3
Formula Weight 143.94
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC193530050

  SDS
thermo fisher scientific
AC193530050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Purity Grade Trace Metal Basis
Formula Weight 143.94
Physical Form Powder
Quantity 5g
Color Light gray-green to green
Assay 64.3 to 69.0%
Fieser 04,203; 13,279; 16,227
CAS Min % 100.0
CAS Max % 100.0
Linear Formula MoO3
Total Trace Metal Impurities 10ppm max.
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement:
Suspected of causing cancer.
May cause respiratory irritation.
Causes serious eye irritation.
Assay Percent Range 99.999%
Molecular Formula MoO3
CAS 1313-27-5
Chemical Name or Material Molybdenum(VI) oxide
Synonym Molybdenum trioxide
Packaging Glass bottle
Health Hazard 3 GHS P Statement:
Use personal protective equipment as required.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

For ultra trace analysis

Safety and Handling

Warning

GHS P Statement:
Use personal protective equipment as required.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Suspected of causing cancer.
May cause respiratory irritation.
Causes serious eye irritation.