Learn More
Molybdenum(VI) oxide, 99.95% (metals basis), Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 011837A3
Description
Molybdenum(VI) oxide is a very promising and stable molybdenum oxide for the electrochromic applications and used as a catalyst in oil industries. It is also used to make metal molybdenum. It has a wide band gap n-type semiconductor with distinctive electrochromic, thermochromic, and photochromic properties. It has been widely investigated as a smart material for catalysis sensors, lubricants, lithium battery, organic solar cells, and display materials. Molybdenum(VI) oxide (MoO3) is employed as an adhesive between enamels and metals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
1155°C (sublimation) | |
795°C | |
White | |
2 kg | |
MoO3 | |
molybdenum trioxide, molybdenum vi oxide, molybdic anhydride, molybdic trioxide, molybdena, molybdic oxide, molybdenum oxide, molybdic acid anhydride, molybdenum peroxide | |
JKQOBWVOAYFWKG-UHFFFAOYSA-N | |
trioxomolybdenum | |
14802 | |
143.94 | |
White to gray powder |
Molybdenum(VI) oxide | |
1313-27-5 | |
Sublimed Powder | |
99.95% (metals basis) | |
MFCD00003469 | |
Barely soluble in water; soluble in base. | |
O=[Mo](=O)=O | |
143.95 | |
CHEBI:30627 | |
Odorless | |
4.692 |
Chemical Identifiers
1313-27-5 | |
143.95 | |
JKQOBWVOAYFWKG-UHFFFAOYSA-N | |
14802 | |
trioxomolybdenum |
MoO3 | |
MFCD00003469 | |
molybdenum trioxide, molybdenum vi oxide, molybdic anhydride, molybdic trioxide, molybdena, molybdic oxide, molybdenum oxide, molybdic acid anhydride, molybdenum peroxide | |
CHEBI:30627 | |
O=[Mo](=O)=O |
Safety and Handling
P201-P202-P261-P264b-P271-P280i-P281-P304+P340-P305+P351+P338-P308+P313-P501c
H319-H335-H351
DOTInformation : DOT Class: 6.1, Packaging Group: III
EINECSNumber : 215-204-7
RTECSNumber : QA4725000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only