Please login to your online account to display your discounted pricing

Alfa Aesar™ Molybdenum(VI) fluoride

Chemical Identifiers

Chemical Name or Material Molybdenum(VI) fluoride
CAS 7783-77-9
Linear Formula MoF6
Formula Weight 209.93
MDL Number MFCD00014220
UN Number UN3289
Merck Index 14,6237
Synonym Molybdenum hexafluoride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA2271318

  SDS
alfa aesar™
2271318
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 209.93
Chemical Name or Material Molybdenum(VI) fluoride
TSCA TSCA
Packaging CGA 330 valve cylinder
Physical Form Liquid
Sensitivity Moisture Sensitive
Linear Formula MoF6
MDL Number MFCD00014220
CAS 7783-77-9
UN Number UN3289
Merck Index 14,6237
Synonym Molybdenum hexafluoride
Boiling Point 37°C
Color Colorless
Melting Point 17°C
Quantity 50g
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H330-H314-H318
Fatal if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3 GHS P Statement
P280-P305+P351+P338-P309-P310-P402+P404
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
Store in a dry place.
Store in a closed container.
Storage Note 1
  • Packed in one-way cylinder w/CGA 330 valve
Density 2.3g/mL
DOT Information Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC LIQUID, CORROSIVE, INORGANIC, N.O.S.
RTECS Number QA4681450

  • Packed in one-way cylinder w/CGA 330 valve

Safety and Handling

Danger

GHS P Statement
P280-P305+P351+P338-P309-P310-P402+P404
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
Store in a dry place.
Store in a closed container.

GHS H Statement
H330-H314-H318
Fatal if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.

  • Packed in one-way cylinder w/CGA 330 valve