missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Molybdenum(V) trichloride-isopropoxide, 25% (w/v) in isopropanol

Manufacturer:  Alfa Aesar™ 44474AD

 View more versions of this product

Catalog No. AA44474AD

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

Molybdenum (V) trichloride-isopropoxide
50mL
C6H16Cl3MoO2
UN2924
25% (w/v) in isopropanol
CC(C)O.CC(C)O.Cl[Mo](Cl)Cl
322.492
320.48
Moisture Sensitive
53780-81-7
MFCD02682829
molybdenum v trichloride-isopropoxide, in isopropanol
JVWWBSIISOXHDE-UHFFFAOYSA-K
propan-2-ol;trichloromolybdenum
121233634
Liquid

Chemical Identifiers

53780-81-7
322.492
JVWWBSIISOXHDE-UHFFFAOYSA-K
121233634
CC(C)O.CC(C)O.Cl[Mo](Cl)Cl
C6H16Cl3MoO2
MFCD02682829
molybdenum v trichloride-isopropoxide, in isopropanol
propan-2-ol;trichloromolybdenum
Safety and Handling

Safety and Handling

GHS H Statement
H225-H314-H318
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P210-P280-P303+P361+P353-P305+P351+P338-P310
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

Danger

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S.

SDS Documents