Please login to your online account to display your discounted pricing

Alfa Aesar™ Molybdenum(V) trichloride-isopropoxide, 25% (w/v) in isopropanol

$110.14 - $110.14

Chemical Identifiers

Chemical Name or Material Molybdenum (V) trichloride-isopropoxide
CAS 53780-81-7
Molecular Formula MoCl3(OC3H7)2
Formula Weight 320.48
MDL Number MFCD02682829
UN Number UN2924
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA44474AB

  SDS
alfa aesar™
44474AB
10mL Each for $110.14
Description & Specifications

Specifications

Formula Weight 320.48
Health Hazard 1 Danger
Health Hazard 3 GHS P Statement
P210-P280-P303+P361+P353-P305+P351+P338-P310
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Chemical Name or Material Molybdenum (V) trichloride-isopropoxide
Sensitivity Moisture Sensitive
CAS 53780-81-7
Health Hazard 2 GHS H Statement
H225-H314-H318
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Molecular Formula MoCl3(OC3H7)2
Quantity 10mL
Concentration 25% (w/v) in isopropanol
MDL Number MFCD02682829
DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S.
Physical Form Liquid
UN Number UN2924
Safety and Handling

Danger

GHS P Statement
P210-P280-P303+P361+P353-P305+P351+P338-P310
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H225-H314-H318
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
Causes serious eye damage.