Learn More
Molybdenum(V) isopropoxide, 99.6% (m.b.), 5% w/v in isopropanol, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 041836AE
Description
Used in the preparations of nanomaterials and thin films.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Packaged under argon
Specifications
Molybdenum(V) isopropoxide | |
Yellow to Brown | |
Alcohol-like | |
(metals basis) | |
Mo[OCH(CH3)2]5 | |
100 mL | |
Moisture sensitive | |
Soluble in hydrocarbons,reacts with alcohols,ketones and esters | |
CC(C)O[Mo](OC(C)C)(OC(C)C)(OC(C)C)OC(C)C | |
391.39 | |
391.04 | |
5% w/v in isopropanol |
209733-38-0 | |
12°C (54°F) | |
ChemSeal™ bottles | |
C15H35MoO5 | |
MFCD00210636 | |
UN1219 | |
molybdenum v isopropoxide, acmc-20akdz, pentaisopropoxymolybdenum v, molybdenum 5+ pentakis propan-2-olate | |
HLFWWCCBFJUYJL-UHFFFAOYSA-N | |
molybdenum;propan-2-olate | |
18475294 | |
99.6% | |
Liquid |
Chemical Identifiers
209733-38-0 | |
391.39 | |
HLFWWCCBFJUYJL-UHFFFAOYSA-N | |
18475294 | |
CC(C)O[Mo](OC(C)C)(OC(C)C)(OC(C)C)OC(C)C |
C15H35MoO5 | |
MFCD00210636 | |
molybdenum v isopropoxide, acmc-20akdz, pentaisopropoxymolybdenum v, molybdenum 5+ pentakis propan-2-olate | |
molybdenum;propan-2-olate |
Safety and Handling
GHS H Statement
H225-H315-H319-H336
Highly flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause drowsiness or dizziness.
P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P370+P378q-P501c
H225-H319-H335-H336
DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ISOPROPANOL
TSCA : No
storageNote1 :
- Packaged under argon
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only