missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Molybdenum(V) chloride, 99.6% (metals basis), Alfa Aesar™

Manufacturer:  Alfa Aesar 1183230

 View more versions of this product

Catalog No. AA1183230



Specifications

Specifications

Molybdenum(V) chloride
99.6% (metals basis)
MFCD00003467
acmc-1bx8l
Cl[Mo](Cl)(Cl)(Cl)Cl
273.2
CHEBI:30635
Crystalline
2.3 hPa (2mm Hg) at 25°C
250g
10241-05-1
Cl5Mo
Black crystals
GICWIDZXWJGTCI-UHFFFAOYSA-I
pentachloromolybdenum
61497
273.24
Pungent
White to Gray

Chemical Identifiers

10241-05-1
273.2
GICWIDZXWJGTCI-UHFFFAOYSA-I
61497
pentachloromolybdenum
Cl5Mo
MFCD00003467
acmc-1bx8l
CHEBI:30635
Cl[Mo](Cl)(Cl)(Cl)Cl
Safety and Handling

Safety and Handling

H314-H318-H351: Causes severe skin burns and eye damage. Causes serious eye damage. Suspected of causing cancer.

DOTInformation : DOT Class: 8, Packaging Group: III

EINECSNumber : 233-575-3

Documents