missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Molybdenum(V) chloride, 95%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 197081000

 View more versions of this product

Catalog No. AC197081000



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

C-X bond formation reagent
Specifications

Specifications

268°C
194°C
10241-05-1
94.0
Fine Crystals or Crystalline Powder
94% min. (Argentometry)
MoCl5
2508
molybdenum v chloride, molybdenum pentachloride, molybdenum chloride mocl5, mocl5, pentachloridomolybdenum, molybdenum 5+ chloride, acmc-1bx8l, ksc491a7h
GICWIDZXWJGTCI-UHFFFAOYSA-I
pentachloromolybdenum
61497
273.2
Glass bottle
Molybdenum(V) chloride
98%
Black to Blue or Black-Green
100.0
100 g
Cl5Mo
MFCD00003467
06,412; 01,1015; 02,365; 03,250; 04,428; 05,587; 06,519; 08,443; 09,414; 11,471; 12,434; 13,273; 04,99; 16,95
Solubility in water: reacts with water. Other solubilities: soluble in dry ether,dry alcohol and other,organic solvents
[Cl-].[Cl-].[Cl-].[Cl-].[Cl-].[Mo+5]
273.20
CHEBI:30635
95%

Chemical Identifiers

10241-05-1
273.20
GICWIDZXWJGTCI-UHFFFAOYSA-I
61497
[Cl-].[Cl-].[Cl-].[Cl-].[Cl-].[Mo+5]
Cl5Mo
MFCD00003467
molybdenum v chloride, molybdenum pentachloride, molybdenum chloride mocl5, mocl5, pentachloridomolybdenum, molybdenum 5+ chloride, acmc-1bx8l, ksc491a7h
CHEBI:30635
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Causes severe skin burns and eye damage.
Reacts violently with water.
Contact with water liberates toxic gas.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi

GHS Signal Word: Danger

EINECSNumber : 233-575-3

RTECSNumber : QA4690000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only