Please login to your online account to display your discounted pricing

Molybdenum Standard For ICP, 1000μg/mL in H2O, SPEX CertiPrep™

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

$83.09 - $305.20

Chemical Identifiers

Chemical Name or Material Molybdenum Standard
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
View More Specs
Includes:  NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

 Disclaimers

Product intended for laboratory use only

Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

PLMO92T

 
spex certiprep llc
PLMO92T
Poly Bottle 250mL Each for $247.06

PLMO9-2X

 
spex certiprep llc
PLMO92X
Triple-leached LDPE bottle 500mL Each for $305.20

PLMO9-2Y

 
spex certiprep llc
PLMO92Y
Triple-leached LDPE bottle 125mL Each for $174.40

PLMO92M

 
spex certiprep llc
SPXPLMO92M
Triple-leached LDPE bottle 30mL Each for $83.09
Description & Specifications

Specifications

Quantity 250mL
Health Hazard 1 Harmful in contact with skin. Causes serious eye irritation
Recommended Storage Ambient
DOT Information Nonhazardous
Purity Grade Assurance Grade
Certificate of Analysis Assurance Certificate of Analysis reports up to 30 elements at ppb levels.
CAS 7439-98-7
Packaging Poly Bottle
CAS Max % 99.9
Chemical Name or Material Molybdenum Standard
CAS Min % 0.1
CAS Max % 0.1
CAS 7732-18-5
CAS Min % 99.9

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

AnalyteConcentration
Mo1000μg/mL

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

GHS/OSHA Complaint

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Safety and Handling

Harmful in contact with skin. Causes serious eye irritation

Ambient