Please login to your online account to display your discounted pricing

Alfa Aesar™ Molybdenum sputtering target, 76.2mm (3.0 in.) dia. x 6.35mm (0.250 in.) thick, 99.95% (metals basis)

$300.16 - $300.16

Chemical Identifiers

Chemical Name or Material Molybdenum sputtering target
CAS 7439-98-7
Name Note 76.2mm (3.0 in.) dia. x 6.35mm (0.250 in.) thick
MDL Number MFCD00003465
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA40869KS

  SDS
alfa aesar™
40869KS
1 each Each for $300.16
Description & Specifications

Specifications

Chemical Name or Material Molybdenum sputtering target
TSCA TSCA
Name Note 76.2mm (3.0 in.) dia. x 6.35mm (0.250 in.) thick
Assay 99.95%
Assay Percent Notes (metals basis)
MDL Number MFCD00003465
CAS 7439-98-7
Quantity 1 each