missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Molybdenum sputtering target, 50.8mm (2.0 in.) dia. x 6.35mm (0.250 in.) thick, 99.95% (metals basis)

$266.27 - $266.27

Chemical Identifiers

CAS 7439-98-7
MDL Number MFCD00003465
Chemical Name or Material Molybdenum sputtering target, 50.8mm (2.0 in.) dia. x 6.35mm (0.250 in.) thick
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA40867KS SDS Alfa Aesar™
40867KS
1 each Each for $266.27
Specifications Description & Specifications

Chemical Identifiers

7439-98-7
MFCD00003465
Molybdenum sputtering target, 50.8mm (2.0 in.) dia. x 6.35mm (0.250 in.) thick

Specifications

1 each
MFCD00003465
99.95%, (metals basis)
7439-98-7
Molybdenum sputtering target, 50.8mm (2.0 in.) dia. x 6.35mm (0.250 in.) thick

Safety and Handling

TSCA : TSCA