Please login to your online account to display your discounted pricing

Alfa Aesar™ Molybdenum sputtering target, 50.8mm (2.0 in.) dia. x 3.18mm (0.125 in.) thick, 99.95% (metals basis)

Magnetron sputtering source

$512.30 - $512.30

Chemical Identifiers

Chemical Name or Material Molybdenum sputtering target
CAS 7439-98-7
Name Note 50.8mm (2.0 in.) dia. x 3.18mm (0.125 in.) thick
MDL Number MFCD00003465
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA40866KS

  SDS
alfa aesar™
40866KS
1 each Each for $512.30
Description & Specifications

Specifications

Chemical Name or Material Molybdenum sputtering target
TSCA TSCA
Name Note 50.8mm (2.0 in.) dia. x 3.18mm (0.125 in.) thick
Assay 99.95%
Assay Percent Notes (metals basis)
MDL Number MFCD00003465
CAS 7439-98-7
Quantity 1 each