Please login to your online account to display your discounted pricing

Alfa Aesar™ Molybdenum silicide sputtering target, 76.2mm (3.0 in.) dia. x 6.35mm (0.250 in.) thick, 99.5% (metals basis)

$986.93 - $986.93

Chemical Identifiers

Chemical Name or Material Molybdenum silicide sputtering target
CAS 12136-78-6
Name Note 76.2mm (3.0 in.) dia. x 6.35mm (0.250 in.) thick
Formula Weight 152.13
MDL Number MFCD00151535
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA41079KS

  SDS
alfa aesar™
41079KS
1 each Each for $986.93
Description & Specifications

Specifications

Formula Weight 152.13
Melting Point 1870° to 2030°C
Quantity 1 each
Color Dark gray
Chemical Name or Material Molybdenum silicide sputtering target
Assay Percent Notes (metals basis)
TSCA TSCA
CAS 12136-78-6
Physical Form Target
Assay 99.5%
Density 6.31g/mL
Odor Odorless
Name Note 76.2mm (3.0 in.) dia. x 6.35mm (0.250 in.) thick
MDL Number MFCD00151535